Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018404.2 | NP_060874.1 | 3, 4 | 596 | ||
GenBank mRNA | AJ238994.1 | - | 3, 4 | 605 | ||
AJ242782.1 | - | 3, 4 | 610 | |||
AJ272195.1 | - | 3, 4 | 417 | |||
AK074204.1 | - | 3, 4 | 424 | |||
AK222526.1 | - | 3, 4 | 424 | |||
BC033758.1 | - | 3, 4 | 412 | 5214358 | ||
DQ288859.1 | - | 3, 4 | 418 |