Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018449.3 | NP_060919.3 | 2 | 185 | ||
GenBank mRNA | AB209911.1 | - | 2 | 161 | ||
AF157327.1 | - | 1 | 98 | |||
AK294952.1 | - | 2 | 176 | |||
AK299183.1 | - | 2 | 176 | |||
AK302335.1 | - | 2 | 165 | |||
AK302754.1 | - | 2 | 174 | |||
BG723831.1 | - | 2 | 191 | |||
CR933678.1 | - | 2 | 176 |