Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282529.1 | NP_001269458.1 | 9 | 992 | ||
NM_018449.3 | NP_060919.3 | 13 | 1306 | |||
GenBank mRNA | AB040924.1 | - | 1 | 108 | ||
AK025801.1 | - | 2 | 196 | |||
AK026088.1 | - | 5 | 474 | |||
AK294952.1 | - | 13 | 1224 | |||
AK299183.1 | - | 7 | 714 | |||
AK302335.1 | - | 11 | 1127 | |||
AK302754.1 | - | 12 | 1174 | |||
AK303493.1 | - | 9 | 930 | |||
AY358682.1 | - | 1 | 516 | |||
BC111762.1 | - | 7 | 676 |