Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018460.3 | NP_060930.3 | 9 | 896 | ||
XM_011511479.2 | XP_011509781.1 | 10 | 928 | |||
XM_011511481.2 | XP_011509783.1 | 9 | 831 | |||
XM_011511482.2 | XP_011509784.1 | 9 | 831 | |||
XM_011511483.1 | XP_011509785.1 | 5 | 526 | |||
XM_011511484.1 | XP_011509786.1 | 9 | 831 | |||
XM_017004499.1 | XP_016859988.1 | 9 | 857 | |||
XM_017004500.1 | XP_016859989.1 | 8 | 723 | |||
XM_017004501.1 | XP_016859990.1 | 9 | 831 | |||
GenBank mRNA | AF212222.1 | - | 9 | 836 | ||
AF217507.1 | - | 9 | 837 | |||
AF217522.1 | - | 9 | 834 | |||
AK226014.1 | - | 9 | 831 | |||
AK308219.1 | - | 9 | 854 | |||
AY219338.1 | - | 9 | 834 | |||
BC016701.2 | - | 9 | 802 | 4108644 | ||
BC038976.1 | - | 9 | 801 | 5553184 |