Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130182.1 | NP_001123654.1 | 6 | 1080 | ||
NM_001130183.1 | NP_001123655.1 | 6 | 907 | |||
NM_018602.3 | NP_061072.3 | 7 | 1219 | |||
XM_005254535.3 | XP_005254592.1 | 7 | 992 | |||
XM_006720601.1 | XP_006720664.1 | 7 | 994 | |||
GenBank mRNA | AF173388.1 | - | 2 | 818 | ||
AK025069.1 | - | 1 | 202 | |||
AK090898.1 | - | 3 | 551 | |||
AK096616.1 | - | 6 | 1080 | |||
AK098079.1 | - | 7 | 1210 | |||
AL133096.1 | - | 1 | 992 | |||
AL133433.1 | - | 1 | 321 | |||
AL133434.1 | - | 1 | 418 | |||
BC021720.2 | - | 7 | 1219 | 4831235 | ||
BC031044.1 | - | 6 | 942 | 5277834 | ||
BX648710.1 | - | 7 | 1101 |