Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018654.1 | NP_061124.1 | 1 | 466 | ||
XM_005253421.3 | XP_005253478.1 | 3 | 2906 | |||
XM_017019582.1 | XP_016875071.1 | 2 | 999 | |||
XM_017019583.1 | XP_016875072.1 | 2 | 998 | |||
GenBank mRNA | AB099817.1 | - | 1 | 466 | ||
AF209923.1 | - | 1 | 466 | |||
BC069341.1 | - | 1 | 466 | 7262221 | ||
BC107077.1 | - | 1 | 466 | 40027061 | ||
BC107078.1 | - | 1 | 466 | 40027066 |
Set Membership: |
G-protein coupled receptor |