Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018685.4 | NP_061155.2 | 8, 9 | 1754 | ||
XM_006715746.1 | XP_006715809.1 | 8, 9 | 1751 | |||
XM_017012354.1 | XP_016867843.1 | 8, 9 | 1751 | |||
XM_017012355.1 | XP_016867844.1 | 8, 9 | 1751 | |||
GenBank mRNA | AF273437.1 | - | 8, 9 | 1689 | ||
AK299803.1 | - | 8, 9 | 1456 | |||
CR936650.1 | - | 5, 6 | 844 |