Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018925.2 | NP_061748.1 | 1 | 737 | ||
NM_032099.1 | NP_115270.1 | 1 | 737 | |||
GenBank mRNA | AF152334.1 | - | 1 | 737 | ||
AF152521.1 | - | 1 | 737 | |||
BC093877.1 | - | 1 | 828 | 7939722 | ||
BC093879.1 | - | 1 | 828 | 7939724 |