Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018936.3 | NP_061759.1 | 1 | 285 | ||
GenBank mRNA | AB209745.1 | - | 1 | 61 | ||
AF152495.1 | - | 1 | 101 | |||
AF217756.1 | - | 1 | 101 | |||
AK027526.1 | - | 1 | 257 | |||
BC098347.1 | - | 1 | 148 | 40007951 | ||
BC098575.1 | - | 1 | 249 | 30921306 | ||
DA703023.1 | - | 1 | 285 |