Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287813.2 | NP_001274742.1 | 12 | 1057 | ||
NM_001287814.2 | NP_001274743.1 | 10 | 1209 | |||
NM_001321773.1 | NP_001308702.1 | 10 | 1209 | |||
NM_001321775.1 | NP_001308704.1 | 12 | 1057 | |||
NM_001321776.1 | NP_001308705.1 | 11 | 1284 | |||
NM_001321777.1 | NP_001308706.1 | 10 | 1209 | |||
NM_001321778.1 | NP_001308707.1 | 11 | 1015 | |||
NM_018967.4 | NP_061840.1 | 11 | 1284 | |||
NR_135794.1 | 11 | 1264 | ||||
NR_135795.1 | 11 | 1284 | ||||
NR_135796.1 | 11 | 1284 | ||||
NR_135797.1 | 11 | 1264 | ||||
NR_135798.1 | 10 | 1209 | ||||
XM_011517548.2 | XP_011515850.1 | 10 | 1394 | |||
XM_017013579.1 | XP_016869068.1 | 13 | 4213 | |||
XM_017013580.1 | XP_016869069.1 | 11 | 807 | |||
XM_017013581.1 | XP_016869070.1 | 11 | 1529 | |||
XM_017013582.1 | XP_016869071.1 | 11 | 1529 | |||
GenBank mRNA | AJ003030.1 | - | 10 | 845 | ||
AK314084.1 | - | 12 | 994 | |||
AL161971.1 | - | 11 | 1179 | |||
BC075072.2 | - | 9 | 649 | 30915592 | ||
BC104829.1 | - | 10 | 731 | 8143832 | ||
BC143695.1 | - | 10 | 731 |