Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018968.3 | NP_061841.2 | 3, 4 | 404 | ||
XM_017004361.1 | XP_016859850.1 | 3, 4 | 366 | |||
XM_017004362.1 | XP_016859851.1 | 3, 4 | 372 | |||
XM_017004363.1 | XP_016859852.1 | 3, 4 | 553 | |||
XM_017004365.1 | XP_016859854.1 | 3, 4 | 367 | |||
XM_017004366.1 | XP_016859855.1 | 3, 4 | 367 | |||
XM_017004367.1 | XP_016859856.1 | 3, 4 | 404 | |||
XM_017004368.1 | XP_016859857.1 | 3, 4 | 367 | |||
XM_017004369.1 | XP_016859858.1 | 3, 4 | 367 | |||
XM_017004370.1 | XP_016859859.1 | 3, 4 | 367 | |||
XM_017004371.1 | XP_016859860.1 | 3, 4 | 368 | |||
XM_017004372.1 | XP_016859861.1 | 3, 4 | 368 | |||
XM_017004374.1 | XP_016859863.1 | 3, 4 | 373 | |||
GenBank mRNA | AJ003029.1 | - | 3, 4 | 406 |