Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271581.1 | NP_001258510.1 | 17 | 2217 | ||
NM_018981.2 | NP_061854.1 | 18 | 2355 | |||
NR_073365.1 | 18 | 3109 | ||||
NR_073366.1 | 19 | 3212 | ||||
NR_073367.1 | 17 | 2248 | ||||
GenBank mRNA | AF038503.1 | - | 18 | 2066 | ||
AF490904.1 | - | 17 | 1965 | |||
AK027450.1 | - | 16 | 1696 | |||
AK027647.1 | - | 18 | 2124 | |||
AK027696.1 | - | 18 | 2103 | |||
AK027701.2 | - | 15 | 1477 | |||
AK074905.1 | - | 5 | 471 | |||
AK298138.1 | - | 13 | 1202 | |||
AL137648.1 | - | 7 | 905 | |||
AL832267.1 | - | 11 | 1055 | |||
AL832632.2 | - | 18 | 2880 | |||
AL832646.3 | - | 16 | 1706 | |||
AY089971.1 | - | 15 | 1602 | |||
AY358577.1 | - | 17 | 1974 | |||
BC070034.1 | - | 19 | 2952 | |||
BC117299.1 | - | 18 | 2070 | 40125850 | ||
BC126168.1 | - | 18 | 2070 | 8991884 |