Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019010.2 | NP_061883.1 | 1 | 294 | ||
GenBank mRNA | AK312744.1 | - | 1 | 293 | ||
BC031559.1 | - | 1 | 278 | 5189289 | ||
DA426874.1 | - | 1 | 293 |