Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018424.3 | NP_060894.2 | 11 | 1545 | ||
NM_019114.4 | NP_061987.3 | 11 | 1545 | |||
XM_011518790.1 | XP_011517092.1 | 11 | 1042 | |||
XM_011518791.2 | XP_011517093.1 | 11 | 1027 | |||
XM_011518792.2 | XP_011517094.1 | 11 | 1027 | |||
XM_011518794.2 | XP_011517096.1 | 11 | 1027 | |||
XM_017014813.1 | XP_016870302.1 | 11 | 1027 | |||
XM_017014814.1 | XP_016870303.1 | 11 | 1027 | |||
XM_017014815.1 | XP_016870304.1 | 11 | 1027 | |||
GenBank mRNA | AB032179.3 | - | 11 | 1545 | ||
AB209187.1 | - | 3 | 137 | |||
AF153416.1 | - | 11 | 1317 | |||
AF153418.1 | - | 11 | 1317 | |||
AK000388.1 | - | 11 | 929 |