Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KIAA1217 SIRNA Assays ›
Gene Symbol:
KIAA1217
Gene Name:
KIAA1217
Gene Aliases:
SKT
Chromosome Location:
Chr.10: 23694746 - 24547848 on Build GRCh38
UniGene ID:
Hs.445885
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132875
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KIAA1217
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098500.2
NP_001091970.1
7
1871
NM_001282767.1
NP_001269696.1
6
1705
NM_001282768.1
NP_001269697.1
6
1705
NM_001282769.1
NP_001269698.1
2
788
NM_001282770.1
NP_001269699.1
2
742
NM_001321681.1
NP_001308610.1
2
788
NM_019590.4
NP_062536.2
6
1705
XM_005252516.3
XP_005252573.1
2
716
XM_011519552.2
XP_011517854.2
7
1760
XM_011519555.2
XP_011517857.2
6
1678
XM_011519558.2
XP_011517860.2
7
1762
XM_011519559.2
XP_011517861.2
6
1452
XM_011519562.2
XP_011517864.2
7
1760
XM_011519564.2
XP_011517866.1
6
1432
XM_011519565.2
XP_011517867.2
6
1451
XM_011519566.2
XP_011517868.2
6
1679
XM_017016416.1
XP_016871905.1
7
1786
XM_017016417.1
XP_016871906.1
6
1705
XM_017016418.1
XP_016871907.1
5
2422
XM_017016419.1
XP_016871908.1
5
2422
XM_017016420.1
XP_016871909.1
5
1169
XM_017016421.1
XP_016871910.1
7
1379
XM_017016422.1
XP_016871911.1
5
1169
XM_017016423.1
XP_016871912.1
2
716
XM_017016424.1
XP_016871913.1
2
788
XM_017016425.1
XP_016871914.1
7
1762
XM_017016426.1
XP_016871915.1
7
1766
XM_017016427.1
XP_016871916.1
7
1532
XM_017016428.1
XP_016871917.1
4
948
XM_017016429.1
XP_016871918.1
8
1920
GenBank mRNA
AB033043.2
-
7
1871
AB291616.1
-
5
1232
AK091142.1
-
6
1705
AK096446.1
-
5
1093
AK300422.1
-
2
727
AK303905.1
-
5
1217
AL833280.1
-
6
1332
BC018764.1
-
2
556
BC098577.1
-
2
696
BC136521.1
-
6
1323
9020511
BC144227.1
-
6
1323
9052747
BX640796.1
-
2
717
BX648451.1
-
2
717
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
embryonic skeletal system development
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0