Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019592.6 | NP_062538.5 | 20 | 3922 | ||
XM_011518862.1 | XP_011517164.1 | 20 | 4199 | |||
GenBank mRNA | AF265230.1 | - | 20 | 3920 | ||
AK002051.1 | - | 8 | 2022 | |||
AK022532.1 | - | 20 | 3918 | |||
AL832910.1 | - | 6 | 1761 | |||
AL834272.1 | - | 10 | 2474 |