Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PARD3 SIRNA Assays ›
Gene Symbol:
PARD3
Gene Name:
par-3 family cell polarity regulator
Gene Aliases:
ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
Chromosome Location:
Chr.10: 34109560 - 34815325 on Build GRCh38
UniGene ID:
Hs.131489
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
132894
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PARD3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001184785.1
NP_001171714.1
4
830
NM_001184786.1
NP_001171715.1
4
830
NM_001184787.1
NP_001171716.1
4
830
NM_001184788.1
NP_001171717.1
4
830
NM_001184789.1
NP_001171718.1
4
830
NM_001184790.1
NP_001171719.1
4
830
NM_001184791.1
NP_001171720.1
4
830
NM_001184792.1
NP_001171721.1
4
830
NM_001184793.1
NP_001171722.1
4
830
NM_001184794.1
NP_001171723.1
4
830
NM_019619.3
NP_062565.2
4
830
XM_005252528.4
XP_005252585.1
4
821
XM_005252530.4
XP_005252587.1
4
821
XM_005252531.4
XP_005252588.1
4
821
XM_005252532.4
XP_005252589.1
4
607
XM_005252534.4
XP_005252591.1
4
821
XM_005252535.4
XP_005252592.1
4
821
XM_005252536.4
XP_005252593.1
4
821
XM_011519569.2
XP_011517871.1
4
821
XM_011519570.2
XP_011517872.1
4
821
XM_011519571.2
XP_011517873.1
4
821
XM_011519572.2
XP_011517874.1
4
821
XM_011519573.2
XP_011517875.1
4
821
XM_011519575.2
XP_011517877.1
3
505
XM_011519576.2
XP_011517878.1
4
608
XM_011519577.2
XP_011517879.1
4
821
XM_011519578.2
XP_011517880.1
4
825
XM_011519579.2
XP_011517881.1
3
506
XM_011519581.2
XP_011517883.1
4
821
XM_011519583.2
XP_011517885.1
4
821
XM_011519584.2
XP_011517886.1
4
821
XM_017016430.1
XP_016871919.1
4
821
GenBank mRNA
AB073671.1
-
4
500
AF196185.1
-
4
787
AF196186.1
-
4
787
AF252293.1
-
4
500
AF332592.1
-
4
500
AF332593.1
-
4
500
AF454057.1
-
3
339
AF454058.1
-
4
844
AF467002.1
-
4
500
AF467003.1
-
4
500
AF467004.1
-
4
500
AF467005.1
-
4
500
AF467006.1
-
4
500
AK027735.1
-
2
127
BC071566.1
-
4
777
30344732
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
protein targeting to membrane
cell cycle
establishment or maintenance of cell polarity
transforming growth factor beta receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
axonogenesis
asymmetric cell division
negative regulation of peptidyl-threonine phosphorylation
myelination in peripheral nervous system
positive regulation of myelination
regulation of cellular localization
bicellular tight junction assembly
establishment of epithelial cell polarity
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
protein phosphatase binding
phosphatidylinositol-3-phosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0