Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4ENIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164501.1 | NP_001157973.1 | 9 | 1419 | ||
NM_001164502.1 | NP_001157974.1 | 7 | 954 | |||
NM_019843.3 | NP_062817.2 | 9 | 1443 | |||
XM_005261686.2 | XP_005261743.1 | 9 | 1443 | |||
XM_005261687.2 | XP_005261744.1 | 9 | 1424 | |||
XM_005261688.2 | XP_005261745.1 | 9 | 1294 | |||
XM_006724281.3 | XP_006724344.3 | 9 | 1720 | |||
XM_011530280.1 | XP_011528582.1 | 9 | 1294 | |||
XM_011530281.2 | XP_011528583.1 | 9 | 1515 | |||
XM_017028863.1 | XP_016884352.1 | 9 | 1720 | |||
XM_017028864.1 | XP_016884353.1 | 9 | 1515 | |||
GenBank mRNA | AF240775.1 | - | 9 | 1273 | ||
AK025254.1 | - | 2 | 171 | |||
AK299136.1 | - | 2 | 270 | |||
AK314636.1 | - | 9 | 1444 | |||
BC012734.2 | - | 9 | 1296 | |||
BC032941.1 | - | 7 | 895 | 5268671 | ||
BC033028.1 | - | 9 | 1393 | 4837225 | ||
CR456386.1 | - | 8 | 1252 |