Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142648.1 | NP_001136120.1 | 8 | 1473 | ||
NM_020150.4 | NP_064535.1 | 7 | 1403 | |||
GenBank mRNA | AF318066.1 | - | 6 | 1268 | ||
AL136724.1 | - | 7 | 1387 | |||
AY008268.1 | - | 7 | 1394 | |||
DR763562.1 | - | 1 | 384 |