Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020199.2 | NP_064584.1 | 2 | 309 | ||
GenBank mRNA | AF226055.1 | - | 2 | 382 | ||
AK074963.1 | - | 2 | 221 | |||
AK298515.1 | - | 2 | 201 | |||
AK315362.1 | - | 2 | 207 | |||
AY157581.1 | - | 2 | 218 | |||
BC020875.1 | - | 2 | 200 | 4610607 | ||
BP245947.1 | - | 2 | 309 |