Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020223.3 | NP_064608.2 | 10 | 2342 | ||
XM_017012450.1 | XP_016867939.1 | 11 | 2598 | |||
XM_017012451.1 | XP_016867940.1 | 11 | 2595 | |||
XM_017012455.1 | XP_016867944.1 | 9 | 5181 | |||
GenBank mRNA | AB545605.1 | - | 10 | 2342 | ||
AK054704.1 | - | 7 | 1212 | |||
AL390147.1 | - | 7 | 1768 | |||
BC040074.1 | - | 10 | 2198 | 4942737 | ||
BC087853.1 | - | 7 | 1233 |