Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020338.3 | NP_065071.1 | 11, 12 | 1525 | ||
XM_011539980.2 | XP_011538282.1 | 5, 6 | 793 | |||
XM_017016442.1 | XP_016871931.1 | 5, 6 | 965 | |||
GenBank mRNA | AB033050.2 | - | 5, 6 | 744 | ||
AY235683.1 | - | 8, 9 | 1022 | |||
BC007288.1 | - | 11, 12 | 1525 |