Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286521.1 | NP_001273450.1 | 5 | 628 | ||
NM_001286522.1 | NP_001273451.1 | 6 | 750 | |||
NM_020367.5 | NP_065100.2 | 6 | 749 | |||
NR_104461.1 | 6 | 749 | ||||
XM_005253711.4 | XP_005253768.1 | 6 | 1143 | |||
XM_011520970.1 | XP_011519272.1 | 7 | 865 | |||
XM_011520971.2 | XP_011519273.1 | 7 | 1265 | |||
XM_011520973.2 | XP_011519275.1 | 6 | 578 | |||
XM_017019670.1 | XP_016875159.1 | 7 | 787 | |||
XM_017019671.1 | XP_016875160.1 | 6 | 1144 | |||
GenBank mRNA | AF263540.1 | - | 5 | 564 | ||
AK299372.1 | - | 5 | 481 | |||
AK314916.1 | - | 6 | 650 | |||
BC017569.1 | - | 6 | 675 | 4077842 | ||
BC031073.1 | - | 6 | 737 | 5284275 | ||
BX640960.1 | - | 5 | 552 | |||
BX647487.1 | - | 6 | 668 | |||
CR749294.1 | - | 6 | 656 |