Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304811.1 | NP_001291740.1 | 5 | 713 | ||
NM_020374.3 | NP_065107.1 | 5 | 721 | |||
XM_005253716.1 | XP_005253773.1 | 5 | 721 | |||
XM_006718992.3 | XP_006719055.1 | 5 | 721 | |||
GenBank mRNA | AJ272205.1 | - | 5 | 664 | ||
AK074479.1 | - | 5 | 721 | |||
AL834377.1 | - | 5 | 684 | |||
BC016908.1 | - | 5 | 664 | 4402263 | ||
BX641128.1 | - | 5 | 713 |