Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289010.1 | NP_001275939.1 | 8, 9 | 1595 | ||
NM_020379.3 | NP_065112.1 | 8, 9 | 1595 | |||
XM_017001860.1 | XP_016857349.1 | 7, 8 | 698 | |||
GenBank mRNA | AB209275.1 | - | 8, 9 | 943 | ||
AF261655.1 | - | 8, 9 | 1595 | |||
BC137017.1 | - | 8, 9 | 1401 | 9021010 |