Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020404.2 | NP_065137.1 | 1 | 2109 | ||
GenBank mRNA | AF279142.1 | - | 1 | 2097 | ||
AK027290.1 | - | 1 | 1231 | |||
BC051340.1 | - | 1 | 2096 | 6339237 | ||
BC104484.1 | - | 1 | 2109 | 40007855 | ||
BC105633.1 | - | 1 | 2109 | 40007854 |