Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164841.2 | NP_001158313.1 | 4 | 590 | ||
NM_020408.5 | NP_065141.3 | 3 | 465 | |||
NR_104417.1 | 4 | 812 | ||||
NR_104418.1 | 2 | 344 | ||||
GenBank mRNA | AF088060.1 | - | 3 | 245 | ||
AF170070.1 | - | 3 | 261 | |||
AF285118.1 | - | 3 | 261 | |||
AK291158.1 | - | 3 | 257 | |||
AK309458.1 | - | 3 | 511 | |||
BC009552.2 | - | 3 | 316 | 3898393 | ||
BC067762.1 | - | 2 | 136 | |||
BC105601.1 | - | 2 | 80 | |||
BI226933.1 | - | 3 | 440 | |||
BI758258.1 | - | 3 | 454 | |||
BX353584.2 | - | 4 | 369 | |||
BX446965.2 | - | 1 | 44 | |||
HY168919.1 | - | 3 | 466 |