Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOLPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135917.1 | NP_001129389.1 | 7 | 1448 | ||
NM_020438.4 | NP_065171.2 | 8 | 1577 | |||
NR_072983.1 | 6 | 1363 | ||||
GenBank mRNA | BC009493.2 | - | 8 | 1547 | ||
BC033686.1 | - | 8 | 1555 | 5195127 | ||
BC040152.1 | - | 6 | 1333 |