Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018070.2 | NP_001018080.1 | 4 | 423 | ||
NM_020441.2 | NP_065174.1 | 3 | 401 | |||
GenBank mRNA | AK000860.1 | - | 3 | 384 | ||
AK309267.1 | - | 3 | 376 | |||
AK315399.1 | - | 3 | 356 | |||
BC006449.1 | - | 3 | 351 | 3958930 | ||
BI767141.1 | - | 4 | 399 | |||
DA061767.1 | - | 4 | 423 |