Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127482.2 | NP_001120954.1 | 3 | 562 | ||
NM_020456.3 | NP_065189.1 | 4 | 679 | |||
NR_023351.2 | 2 | 395 | ||||
XM_017020677.1 | XP_016876166.1 | 4 | 679 | |||
GenBank mRNA | AF052154.1 | - | 4 | 478 | ||
AF055016.1 | - | 4 | 478 | |||
AF334405.1 | - | 4 | 518 | |||
AK290576.1 | - | 3 | 401 | |||
AK308818.1 | - | 4 | 512 | |||
AK316576.1 | - | 4 | 485 | |||
BC022519.1 | - | 3 | 368 | 4792366 | ||
DA388141.1 | - | 2 | 220 |