Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020666.2 | NP_065717.1 | 6 | 743 | ||
GenBank mRNA | AF294429.1 | - | 5 | 635 | ||
AK311485.1 | - | 5 | 654 | |||
BC015942.1 | - | 6 | 743 | |||
BC136261.1 | - | 6 | 691 | 9020248 | ||
BC143547.1 | - | 6 | 691 | 9052058 | ||
BC151233.1 | - | 6 | 1957 | |||
BX647323.1 | - | 2 | 600 | |||
CR749504.1 | - | 8 | 1622 |