Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282145.1 | NP_001269074.1 | 7 | 4373 | ||
NM_001282146.1 | NP_001269075.1 | 6 | 4048 | |||
NM_020742.3 | NP_065793.1 | 6 | 3883 | |||
NM_181332.2 | NP_851849.1 | 6 | 4066 | |||
XM_005274564.2 | XP_005274621.1 | 7 | 4120 | |||
XM_005274565.2 | XP_005274622.1 | 7 | 4103 | |||
XM_005274566.4 | XP_005274623.1 | 7 | 4047 | |||
XM_006724504.3 | XP_006724567.1 | 7 | 3988 | |||
XM_011545547.2 | XP_011543849.1 | 7 | 4122 | |||
XM_011545548.2 | XP_011543850.1 | 7 | 4286 | |||
XM_017029690.1 | XP_016885179.1 | 7 | 3932 | |||
XM_017029691.1 | XP_016885180.1 | 6 | 4062 | |||
XM_017029692.1 | XP_016885181.1 | 6 | 3987 | |||
XM_017029693.1 | XP_016885182.1 | 6 | 3872 | |||
GenBank mRNA | AB033086.1 | - | 6 | 3883 | ||
AF376803.1 | - | 6 | 3883 | |||
AX773938.1 | - | 7 | 3994 | |||
BC034018.1 | - | 6 | 3849 | 4702963 |