Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282145.1 | NP_001269074.1 | 7 | 4893 | ||
NM_001282146.1 | NP_001269075.1 | 6 | 4568 | |||
NM_020742.3 | NP_065793.1 | 6 | 4403 | |||
NM_181332.2 | NP_851849.1 | 6 | 4586 | |||
XM_005274564.2 | XP_005274621.1 | 7 | 4640 | |||
XM_005274565.2 | XP_005274622.1 | 7 | 4623 | |||
XM_005274566.4 | XP_005274623.1 | 7 | 4567 | |||
XM_006724504.3 | XP_006724567.1 | 7 | 4508 | |||
XM_011545547.2 | XP_011543849.1 | 7 | 4642 | |||
XM_011545548.2 | XP_011543850.1 | 7 | 4806 | |||
XM_017029690.1 | XP_016885179.1 | 7 | 4452 | |||
XM_017029691.1 | XP_016885180.1 | 6 | 4582 | |||
XM_017029692.1 | XP_016885181.1 | 6 | 4507 | |||
XM_017029693.1 | XP_016885182.1 | 6 | 4392 | |||
GenBank mRNA | AB033086.1 | - | 6 | 4403 | ||
AF376803.1 | - | 6 | 4403 | |||
AX773938.1 | - | 7 | 4514 |