Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292003.1 | NP_001278932.1 | 10 | 2850 | ||
NM_001292006.1 | NP_001278935.1 | 10 | 2579 | |||
NM_001292007.1 | NP_001278936.1 | 9 | 2258 | |||
NM_020803.4 | NP_065854.3 | 10 | 2807 | |||
NR_120447.1 | 8 | 2071 | ||||
GenBank mRNA | AB037799.2 | - | 9 | 2490 | ||
AK128827.1 | - | 10 | 2848 | |||
BC041384.2 | - | 10 | 2757 | 5275946 | ||
BC041901.1 | - | 8 | 1927 | |||
BX538173.1 | - | 10 | 2724 | |||
BX640727.1 | - | 10 | 2717 | |||
BX640744.1 | - | 10 | 2508 | |||
BX647454.1 | - | 10 | 2743 | |||
BX648367.1 | - | 8 | 1775 | |||
CR749265.1 | - | 9 | 2176 |