Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRAMD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136199.2 | NP_001129671.1 | 13 | 1696 | ||
NM_001320034.1 | NP_001306963.1 | 14 | 1717 | |||
NM_001320035.1 | NP_001306964.1 | 13 | 1604 | |||
NM_001320036.1 | NP_001306965.1 | 15 | 1887 | |||
NM_020895.4 | NP_065946.2 | 14 | 1717 | |||
XM_011527149.1 | XP_011525451.1 | 14 | 1714 | |||
XM_011527153.1 | XP_011525455.1 | 15 | 1890 | |||
XM_011527154.2 | XP_011525456.1 | 14 | 1690 | |||
XM_011527155.1 | XP_011525457.1 | 15 | 1968 | |||
XM_011527156.1 | XP_011525458.1 | 13 | 1604 | |||
XM_017027034.1 | XP_016882523.1 | 14 | 1717 | |||
XM_017027035.1 | XP_016882524.1 | 14 | 1834 | |||
GenBank mRNA | AB040966.1 | - | 12 | 1348 | ||
AK026064.1 | - | 5 | 534 | |||
AK074827.1 | - | 11 | 1280 | |||
AK074864.1 | - | 13 | 1606 | |||
AK074914.1 | - | 13 | 1590 | |||
AK097215.1 | - | 15 | 1887 | |||
BC014077.2 | - | 14 | 1705 | 4126671 | ||
BC041618.1 | - | 4 | 1344 |