Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021229.3 | NP_067052.2 | 3 | 1189 | ||
GenBank mRNA | AF119916.3 | - | 4 | 726 | ||
AF278532.1 | - | 3 | 1191 | |||
AF297711.1 | - | 3 | 1041 | |||
AK024691.1 | - | 2 | 265 | |||
AK226070.1 | - | 3 | 1046 | |||
AK290591.1 | - | 3 | 726 | |||
AK315796.1 | - | 3 | 1048 | |||
AL137540.1 | - | 1 | 57 | |||
AL833767.1 | - | 2 | 593 | |||
AY330211.1 | - | 3 | 740 | |||
BC013591.1 | - | 2 | 264 | |||
BC136798.1 | - | 3 | 1164 | 9020788 | ||
BC136799.1 | - | 3 | 1164 | 9020789 |