Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021620.3 | NP_067633.2 | 4 | 2009 | ||
GenBank mRNA | AY004253.1 | - | 4 | 2039 | ||
AY130010.1 | - | 1 | 252 |