Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RXFP1 SIRNA Assays ›
Gene Symbol:
RXFP1
Gene Name:
relaxin/insulin like family peptide receptor 1
Gene Aliases:
LGR7, RXFPR1
Chromosome Location:
Chr.4: 158521711 - 158653372 on Build GRCh38
UniGene ID:
Hs.591686
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133409
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RXFP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253727.1
NP_001240656.1
6
866
NM_001253728.1
NP_001240657.1
5
686
NM_001253729.1
NP_001240658.1
6
785
NM_001253730.1
NP_001240659.1
5
710
NM_001253732.1
NP_001240661.1
7
869
NM_001253733.1
NP_001240662.1
7
869
NM_021634.3
NP_067647.2
6
785
NR_045579.1
7
1474
NR_045580.1
6
838
NR_045581.1
6
809
NR_045582.1
6
890
NR_045583.1
5
728
NR_045584.1
6
785
XM_011532174.1
XP_011530476.1
6
835
XM_011532175.1
XP_011530477.1
6
835
XM_011532176.1
XP_011530478.1
6
757
XM_011532179.2
XP_011530481.1
6
835
XM_017008517.1
XP_016864006.1
6
835
XM_017008518.1
XP_016864007.1
6
757
XM_017008519.1
XP_016864008.1
7
799
XM_017008520.1
XP_016864009.1
6
888
XM_017008521.1
XP_016864010.1
7
718
XM_017008522.1
XP_016864011.1
6
806
XM_017008523.1
XP_016864012.1
6
757
XM_017008524.1
XP_016864013.1
6
757
XM_017008525.1
XP_016864014.1
5
689
XM_017008526.1
XP_016864015.1
2
328
GenBank mRNA
AB208943.1
-
7
607
AF190500.1
-
6
522
AK122647.1
-
8
826
AK294694.1
-
6
630
AK295040.1
-
7
1297
AK300379.1
-
5
581
AK307889.1
-
5
627
AK312470.1
-
7
701
AY899848.1
-
6
522
AY899849.1
-
6
522
AY899850.1
-
5
423
BC112142.1
-
6
604
8327610
BC113617.1
-
6
607
8322669
BC143556.1
-
6
627
BC143558.1
-
6
660
BC143559.1
-
5
532
BX647975.1
-
6
723
BX647980.1
-
6
723
BX647985.1
-
6
752
CR748189.1
-
3
157
DA156504.1
-
1
36
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0