Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021818.3 | NP_068590.1 | 5 | 2485 | ||
XM_011537057.2 | XP_011535359.1 | 6 | 2548 | |||
XM_017021568.1 | XP_016877057.1 | 6 | 2778 | |||
GenBank mRNA | AK023071.1 | - | 5 | 2308 |