Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003897.1 | NP_001003897.1 | 3 | 601 | ||
NM_022077.3 | NP_071360.1 | 4 | 718 | |||
XM_017028006.1 | XP_016883495.1 | 4 | 1776 | |||
XM_017028007.1 | XP_016883496.1 | 3 | 1659 | |||
GenBank mRNA | AK026708.1 | - | 4 | 1318 | ||
BC014672.1 | - | 3 | 579 | 3535633 | ||
BC016822.1 | - | 5 | 942 | 4102182 |