Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 19 | 2899 | ||
XM_005260504.4 | XP_005260561.1 | 19 | 3612 | |||
XM_017028012.1 | XP_016883501.1 | 18 | 2670 | |||
GenBank mRNA | AF395833.1 | - | 19 | 2876 | ||
AK026157.1 | - | 11 | 1414 | |||
AK074214.1 | - | 4 | 534 | |||
AK122963.1 | - | 2 | 1090 | |||
AK296989.1 | - | 16 | 2470 | |||
BC041331.2 | - | 16 | 2114 | |||
BC142981.1 | - | 16 | 2114 |