Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003811.1 | NP_001003811.1 | 27 | 2415 | ||
NM_031276.2 | NP_112566.2 | 26 | 2293 | |||
XM_011530994.1 | XP_011529296.1 | 26 | 2287 | |||
XM_011530996.1 | XP_011529298.1 | 15 | 1433 | |||
XM_017029649.1 | XP_016885138.1 | 26 | 2287 | |||
XM_017029650.1 | XP_016885139.1 | 15 | 1479 | |||
XM_017029651.1 | XP_016885140.1 | 15 | 1433 | |||
XM_017029652.1 | XP_016885141.1 | 13 | 1114 | |||
GenBank mRNA | AF285594.1 | - | 25 | 2198 | ||
AK057391.1 | - | 15 | 1434 | |||
AK057523.1 | - | 27 | 2409 | |||
AK292471.1 | - | 26 | 2287 | |||
BC036016.1 | - | 26 | 2293 | 5270970 | ||
BU929507.1 | - | 7 | 548 |