Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001260504.1 | NP_001247433.1 | 12, 13 | 1992 | ||
NM_001260505.1 | NP_001247434.1 | 12, 13 | 1716 | |||
NM_031414.4 | NP_113602.2 | 12, 13 | 1716 | |||
NM_032944.3 | NP_116562.2 | 12, 13 | 1960 | |||
NR_048542.1 | 12, 13 | 1708 | ||||
XM_006715758.3 | XP_006715821.1 | 10, 11 | 1096 | |||
XM_011515449.2 | XP_011513751.2 | 12, 13 | 1642 | |||
XM_011515450.1 | XP_011513752.1 | 12, 13 | 1963 | |||
XM_011515452.2 | XP_011513754.2 | 10, 11 | 1109 | |||
GenBank mRNA | AF285599.1 | - | 12, 13 | 1612 | ||
AF332194.1 | - | 12, 13 | 1995 | |||
AK057271.1 | - | 12, 13 | 1664 | |||
AK131214.1 | - | 3, 4 | 418 | |||
AK292474.1 | - | 13, 14 | 1608 | |||
AK302689.1 | - | 12, 13 | 1608 | |||
BC033018.1 | - | 12, 13 | 1610 | |||
BC059374.1 | - | 12, 13 | 1623 | 5268051 | ||
DA533364.1 | - | 4, 5 | 494 |