Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00470 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023925.1 | 4 | 377 | |||
NR_023926.1 | 3 | 331 | ||||
NR_110327.1 | 3 | 321 | ||||
GenBank mRNA | AF295725.1 | - | 4 | 377 | ||
AF295726.1 | - | 3 | 292 | |||
AF295727.1 | - | 2 | 80 | |||
AF295728.1 | - | 4 | 197 | |||
AF295729.1 | - | 2 | 80 | |||
AK057505.1 | - | 3 | 353 |