Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013415.1 | NP_001013433.1 | 5 | 744 | ||
NM_018315.4 | NP_060785.2 | 5 | 909 | |||
NM_033632.3 | NP_361014.1 | 6 | 1083 | |||
XM_011532083.1 | XP_011530385.1 | 8 | 2096 | |||
XM_011532084.1 | XP_011530386.1 | 7 | 2046 | |||
XM_011532085.1 | XP_011530387.1 | 6 | 1953 | |||
XM_011532086.1 | XP_011530388.1 | 9 | 2274 | |||
XM_011532087.1 | XP_011530389.1 | 8 | 2224 | |||
XM_011532088.1 | XP_011530390.1 | 6 | 1482 | |||
XM_017008362.1 | XP_016863851.1 | 8 | 2493 | |||
GenBank mRNA | AB209725.1 | - | 5 | 673 | ||
AF383178.1 | - | 5 | 483 | |||
AF411971.1 | - | 5 | 996 | |||
AF411972.1 | - | 5 | 777 | |||
AK001933.1 | - | 5 | 534 | |||
AK223449.1 | - | 5 | 711 | |||
AK294568.1 | - | 4 | 470 | |||
AK299722.1 | - | 4 | 492 | |||
AK315142.1 | - | 5 | 691 | |||
AY008274.1 | - | 5 | 746 | |||
AY033553.1 | - | 5 | 777 | |||
AY049984.1 | - | 5 | 990 | |||
BC037320.2 | - | 5 | 664 | |||
BC117244.1 | - | 6 | 1051 | 40125795 | ||
BC117246.1 | - | 6 | 1051 | 40125797 | ||
BC143944.1 | - | 7 | 1101 | 9052462 | ||
CR749305.1 | - | 6 | 1214 |