Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317009.1 | NP_001303938.1 | 7 | 3857 | ||
NM_053277.2 | NP_444507.1 | 6 | 3803 | |||
GenBank mRNA | AF448438.1 | - | 6 | 3577 | ||
AF448439.1 | - | 7 | 3631 | |||
AK092733.1 | - | 5 | 2112 | |||
BC040196.2 | - | 6 | 2731 |