Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPEAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272037.1 | NP_001258966.1 | 6 | 892 | ||
NM_144991.2 | NP_659428.2 | 5 | 830 | |||
GenBank mRNA | AJ487962.1 | - | 5 | 830 | ||
BC021197.2 | - | 5 | 719 |