Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175748.3 | NP_786924.2 | 7 | 994 | ||
NR_038150.1 | 6 | 860 | ||||
GenBank mRNA | AK001345.1 | - | 4 | 372 | ||
AK097477.1 | - | 7 | 979 | |||
AK315492.1 | - | 7 | 793 | |||
BC015046.1 | - | 7 | 793 | 3908032 | ||
BC017013.1 | - | 6 | 638 | 3883770 | ||
BC051819.1 | - | 7 | 756 | 6171393 | ||
BX248249.1 | - | 7 | 753 |