Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018190.3 | NP_060660.2 | 8 | 1021 | ||
NM_176824.2 | NP_789794.1 | 8 | 1021 | |||
XM_005263106.3 | XP_005263163.1 | 8 | 1047 | |||
XM_011532079.2 | XP_011530381.1 | 8 | 1092 | |||
XM_011532080.2 | XP_011530382.1 | 8 | 1089 | |||
XM_011532081.2 | XP_011530383.1 | 8 | 1092 | |||
XM_017008357.1 | XP_016863846.1 | 8 | 1044 | |||
XM_017008358.1 | XP_016863847.1 | 8 | 1047 | |||
GenBank mRNA | AF521643.1 | - | 8 | 985 | ||
AF521644.1 | - | 8 | 985 | |||
AK001577.1 | - | 8 | 985 | |||
AK308076.1 | - | 8 | 1012 | |||
BC032691.1 | - | 8 | 938 | 5240398 |