Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021810.4 | NP_068582.2 | 6 | 668 | ||
NM_177980.2 | NP_817089.1 | 18 | 2771 | |||
XM_017027998.1 | XP_016883487.1 | 18 | 3438 | |||
XM_017027999.1 | XP_016883488.1 | 17 | 3369 | |||
XM_017028001.1 | XP_016883490.1 | 16 | 3315 | |||
GenBank mRNA | AK055202.1 | - | 18 | 2760 | ||
AK074477.1 | - | 6 | 656 | |||
BC062570.1 | - | 4 | 545 | |||
BC136306.1 | - | 18 | 2754 | 9020293 | ||
BC143931.1 | - | 18 | 2754 |